Synthware Compression Cap with Hole, Kemtech America
Supplier: Kemtech America
RC005 RC009 RC112 RC2440
60005-050EA 1.52 CAD
60005-050 60005-052 60005-054 60005-056
Synthware Compression Cap with Hole, Kemtech America
Closures Bottle Closures
Replacement caps feature a hole for use with thermometers, inlet adapters, or microscale reaction vials.
  • Nylon construction
  • Open top caps
Caps designed for use with hose connection barbs, septums; or thermoemter adapters.