DuPont™ Tyvek® 400 Shirts
Supplier: DuPont
TY303SWHSM005000 TY303SWHMD005000 TY303SWHLG005000 TY303SWHXL005000 TY303SWH2X005000 TY303SWH3X005000 TY303SWH4X005000 TY303SWH5X005000 TY303SWH6X005000 TY303SWH7X005000
CA10010-097CS 1670.64 CAD
CA10010-097 CA10010-099 CA10010-101 CA10010-103 CA10010-105 CA10010-107 10016-936 10016-938 10016-940 10016-942
DuPont™ Tyvek® 400 Shirts
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Open wrists
  • Extends to hips
  • Front snap closure
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Shirt design features front snap closures for easy donning and doffing. Shirts also feature a laydown collar and open wrists. These shirts are typically worn with TY350S WH pants.
Certifications: Thickness meets ASTM D1777.