Unisex Tops and Pants, TECHstyles®, Encompass
Supplier: Techstyles
9661A 9663A 9664 9665 9652 9653 9654 9656 9671 9676 9672 9673
14224-342CS 1306.12 CAD
14224-342 14224-346 14224-348 14224-350 14224-358 14224-360 14224-362 14224-364 14224-366 14226-830 14226-832 14226-836 14226-838 14226-840
Unisex Tops and Pants, TECHstyles®, Encompass
Disposable Garments
Made of a soft, high-quality spunbond polypropylene material, these garments combine excellent strength with the softness and comfort of linen.
  • V-neck top design assures easy donning
  • Scrub pants are available with an elastic or drawstring waist for a comfortable fit
Can be used as work or building suits.