KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
45643 45644 45645 45646 45647 45663 45664 45665 45666 45667
CA82004-002CS 406.43 CAD
CA82004-002 CA82004-004 CA82004-006 CA82004-008 CA82004-010 CA82004-012 CA82004-014 CA82004-016 CA82004-018 CA82004-020
KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls, Kimberly-Clark Professional
Disposable Garments
Coveralls for hazardous material handling and other applications requiring chemical barrier protection
  • For hazardous material handling and chemical barrier protection
  • Durable, lightweight fabric laminated with SARANEX® 23-P film exterior
  • Comfortable cloth-like interior
  • No-seams front design for splash protection
  • Available with bound or taped seams (Taped seams use SARANEX® 23-P film)
  • Zipper front with storm flap
Meet EPA Level B/C criteria for skin protection.
Made from a durable, lightweight fabric laminated with SARANEX® 23-P film on the exterior. Interior has a comfortable, cloth-like feel against skin. For splash protection, coveralls are designed with no seams in front. Available with bound or taped seams. Taped seams are taped with SARANEX® 23-P film. Zipper front with storm flap.