Septa, Fluoropolymer Resin Silicone, Qorpak®
Supplier: Qorpak
CAP-00513 CAP-00515 CAP-00563
11311-628CS 119.46 CAD
11311-628 93001-762 16151-690
Septa, Fluoropolymer Resin Silicone, Qorpak®
Septa
Resist coring and provide great resealability.
Ideal for when multiple injections are required. Septa maintain PTFE resistance until punctured, then have silicone compatibility.