White Plastic Screw Caps, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
240805 241017 240917 240804 241317 241009 241018 240817 240818
66021-252CS 596.69 CAD
66021-252 66021-759 66021-781 66021-840 66021-861 66021-748 66021-760 66021-817 66021-828
White Plastic Screw Caps, WHEATON®
Closures Chromatography Vial Closures
White plastic screw caps fit Wheaton vials (see 66021-679 series) and other glass scintillation vials.
  • Suitable for marking
  • Not autoclavable
  • Available unlined or choose from a variety of liners