Septa, Silicone, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W224163 224168
71001-636CS 1441.01 CAD
71001-636 16171-776 16171-783 16171-936
Septa, Silicone, Wheaton
Septa
Red silicone septa for use with aluminum seals.
Item 224168 is a specially molded septa with a PTFE inset. The sealing surface of butyl and PTFE effect a more positive seal than plain PTFE faced septa.