Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
224183-05 224183-07 224182-01 224182-07 224183-06 224183-01 224187-01 224182-05
16171-826CS 339.78 CAD
16171-826 16171-828 16171-818 16171-821 16171-827 16171-829 16171-830 16171-819
Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
Closures Chromatography Vial Closures
One-piece center disc tears out, exposing top of seal to syringe
Shelf packs of 100.