VWR® Maximum Fluid-Impervious Butterfly-Style Shoe Covers
Supplier: VWR
VWR®
414004-641 414004-647 414004-648 414004-643 414004-642 414004-646
414004-643CS 186.03 CAD
414004-643 414004-642 414004-641 414004-648 414004-647 414004-646
VWR® Maximum Fluid-Impervious Butterfly-Style Shoe Covers
Disposable Garments
VWR® Maximum Protection Fluid-Impervious Butterfly-Style Shoe Covers are the cleanest and most durable within the entire VWR® Protection line of apparel
  • Fluid-impervious material with seamless bottom to repel water
  • Skid resistance on wet and smooth surfaces
  • Excellent particulate barrier
  • Low linting
  • Serged seams
Shoe covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
These nonconductive shoe covers are engineered to optimize barrier protection, cleanliness, and comfort. Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.