VWR® Maximum Fluid-Impervious Ankle-High Boot Covers
Supplier: VWR International
414004-638 414004-639 414004-640
414004-638CS 344.01 CAD
414004-638 414004-639 414004-640
VWR® Maximum Fluid-Impervious Ankle-High Boot Covers
Disposable Garments
These boot covers are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Fluid-impervious material with seamless bottom to repel water
  • Skid resistance on wet and smooth surfaces
  • Excellent particulate barrier
  • Low linting
Engineered to optimize barrier protection, cleanliness, and comfort
Boot covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validatedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Cleanroom
Bacterial filtration efficiency 0.999
Particle shedding (Helmke Drum) Level I
Water resistance Pass
Tensile strength 28.8 (Filling)
Tearing strength 16.5 (Widthwise Yarns)
Weight 60.21 g/m²