VWR® Maximum Protection Gowns made with Impervious Fabric
Supplier: VWR International
414004-469 414004-470 414004-471 414004-681 414004-682
414004-469CS 433.35 CAD
414004-469 414004-470 414004-471 414004-681 414004-682
VWR® Maximum Protection Gowns made with Impervious Fabric
Disposable Garments
These gowns are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Full protection from fluids and light chemical splash
  • Manufactured from fluid-impervious material
  • Lowest level of particle shedding
  • Clean, low-linting fabric
Gowns provide barrier protection against fluids, biologicals, particulates, and light chemical splash
Elastic cuffs and ties at the neck and waist ensure secure closure. Serged seams. Available in white or blue.
Gowns are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.