VWR® 96-Place Microtube Racks
Supplier: VWR International
HS2345BV HS2345FV HS2345CV HS2345DV HS2345EV HS2345AV
82024-484CS 1130.84 CAD
82024-484 82024-486 82024-488 82024-490 82024-492 82024-494
VWR® 96-Place Microtube Racks
Racks Tube Racks
These polypropylene racks can accommodate ninety-six 0.5 mL microcentrifuge tubes on one side or 1.5 to 2.0 mL microcentrifuge tubes on the opposite side in a standard 8x12 format.
  • Polypropylene construction
  • Wells are more visible and tubes are easier to place
Racks feature easy-to-grasp tabs on each end and a handy snap-in-place lid. Grid lines are molded on the side of the rack that accommodates the 0.5 mL tubes and raised rings are molded on the other side to make the wells more visible and tubes easier to replace.
Ordering information: Assorted rack pack 82024-494 includes one each of blue, green, pink, yellow, and orange.