PicoSolutions Alternative Energy Education Lab Standard Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3019-01 PS-3019-01-D1 PS-3019-01-D2 PS-3019-01-D3 PS-3019-01-D4 PS-3019-01-PK
470355-302EA 194.43 CAD
470355-302 470355-330 470355-336 470355-372 470355-332 470355-308
PicoSolutions Alternative Energy Education Lab Standard Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore alternative energy with a DIY hands-on turbine model.
  • 3rd grade to college level tailored activities for different learning stages
  • Includes DC turbines (horizontal and vertical) and a 7.2 V solar panel for a full spectrum of alternative energy exploration
  • Versatile configuration Supporte enables both AC and DC configurations
  • Subjects spanning sustainable energy, math, aerodynamics, geometry
Build and test a wind turbine kit contents includes vertical and horizontal turbine level 1 kits, six hydro paddles, a multimeter, and a 7.2 V 200 mA solarMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Dimensions 17×16×8"