PicoSolutions Vertical Wind Turbine Standard Kit
Supplier: PICOSOLUTIONS GROUP INC SE
PS-3014-01 PS-3014-01-D1 PS-3014-01-D2 PS-3014-01-D3 PS-3014-01-D4 PS-3014-01-PK
470355-312EA 58.32 CAD
470355-312 470355-320 470355-334 470355-384 470355-316 470355-342
PicoSolutions Vertical Wind Turbine Standard Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Explore vertical wind energy with a DIY hands-on tubine model.
  • 6th grade to college level tailored activities for different learning stages
  • Subjects spanning Sustainable energy, math, aerodynamics, geometry
  • Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind the basic model has a DC motor generator and uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine.
Ordering information: Enhanced learning option: Upgrade available for a DESS courseMore Product Information