Accessories for Safety Funnels with Lid, 'MARCO' V2.0
Supplier: SCAT AMERICAS, INC.
106700 307003 307100 307101 307347 317634 318998
76503-024EA 496.23 CAD
76503-024 76948-908 76948-910 76948-912 76948-920 76948-922 76948-924
Accessories for Safety Funnels with Lid, 'MARCO' V2.0
Funnels Safety Funnels
Replacement parts for SCAT 'MARCO' V2.0 safety funnels, which can be closed safely and prevent the danger of ignition.
  • Suitable for all models of the MARCO series
  • Available in conductive and non-conductive versions
Stainless steel sieve to catch small objects such as stirrers from HPLC equipment.