Synthware Allihn Condensers
Supplier: Kemtech America
C251120 C259200 C259300 C251200 C254300 C254400 C255300 C255400
60001-062EA 202.34 CAD
60001-062 60001-064 60001-066 60001-068 60001-070 60001-072 60001-078 60001-080
Synthware Allihn Condensers
Condensers
With joints at the top and bottom.
  • Allihn style condenser
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
Allihn style condenser is perfect for most chemical distillation applications.
Order Now
SPECIFICATIONS
Environmentally Preferable