Synthware Coiled Condensers, Kemtech America
Supplier: Kemtech America
C121100 C121120 C129200 C124200 C124300 C124400
60000-890EA 317.23 CAD
60000-890 60000-892 60000-894 60000-896 60000-898 60000-900
Synthware Coiled Condensers, Kemtech America
Condensers
With joints at top and bottom.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • jJcketed
Coiled condenser is perfect for most chemical distillation applications.