Synthware Distillation Condensers, Vacuum-Jacketed, Kemtech America
Supplier: Kemtech America
C161100 C164250
60000-784EA 386.66 CAD
60000-784 60000-790
Synthware Distillation Condensers, Vacuum-Jacketed, Kemtech America
Condensers
With 12 mm (int.Ø) indentations to support packing material.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
Vacuum jacketed to improve performance.