Spiral Reflux Condensers, Ace Glass Incorporated
Supplier: Ace Glass
6020-02 9270-04 6020-06 6020-08 6020-04 6020-10 6020-12
89053-788EA 753.34 CAD
89053-788 89074-330 89083-332 89083-334 89053-932 89235-620 89235-622
Spiral Reflux Condensers, Ace Glass Incorporated
Condensers
Reflux condenser with spiral condensing tube features both hose barb inlet and outlet connections at the top, on the same side
Condensers are equipped with standard taper inner and outer joints at bottom and top. Length between joints is approximately 80-90mm longer than the coil length. Manufactured from borosilicate glass.