KIMBLE® KONTES® Allihn Condensers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
431000-2430 431000-2440 431000-2420 431000-2425 431000-2930 431000-2920
KT431000-2430EA 502.5 CAD
KT431000-2430 KT431000-2440 KT431000-2420 KT431000-2425 KT431000-2930 10000-502
KIMBLE® KONTES® Allihn Condensers, DWK Life Sciences
Condensers
This borosilicate glass Allihn condenser are used in many refluxing operations.
  • Standard taper drip joint at bottom and a standard taper outer joint at top
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.