Breathable Microtube Membranes, Diversified Biotech
Supplier: Diversified Biotech
BMTM-1000 BMTM-2000
89032-137CS 4402.48 CAD
89032-137 89032-139
Breathable Microtube Membranes, Diversified Biotech
Seals General Purpose Sealing Films
Micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange
Breathe-EASY polymer-type membranes offer a water vapor transmission rate of 0.7kg/m2/24 hours. Breathe-EASIER membranes are made of white nonwoven rayon and transmit water vapor six times faster than Breathe-EASY membranes for accelerated growth. Both membranes are gamma irradiated to ensure sterility and are free of cytotoxins.
Formatted with an easy-to-use finger tab, membranes are precut to fit a variety of tube sizes. Microtube cap closures will not bind in centrifuge rotors. Convenient scoring makes liner removal quick and easy.