Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
Supplier: Globe Scientific
118127G 118127Y 118127R 118127B 118127C
60820-142PK 84.54 CAD
60820-142 60820-144 60820-146 60820-148 60820-140
Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
Closures Test/Sample Tube Closures
Polyethylene flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes
Closures fit 12mm tubes, and are available in a variety of colors to simplify identification and selection.
They feature two flexible flanges that press firmly against the inside of the tube wall, thereby providing better leak-resistance than standard plug caps.
Order Now
SPECIFICATIONS
Environmentally Preferable