Phenolic Screw Caps with Rubber Liner, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
73800-13415 73800-15415 73800-18415
89003-552CS 402.63 CAD
89003-552 89003-554 89003-556
Phenolic Screw Caps with Rubber Liner, Kimble Chase
Closures Test/Sample Tube Closures
Phenolic cap material and cemented-in rubber liner are resistant to the effects of repeated autoclaving
Suitable for use with 89003-568 series disposable culture tubes and 21020-607 series disposable centrifuge tubes. Designed from ASTM Specification E982.
Order Now
SPECIFICATIONS
Environmentally Preferable