Heavyweight Metalguard Sleeves, Wells Lamont®
Supplier: Wells Lamont
SK-10-KSC SK-14-KSC SK-18-KSC SK-22-KSC SK-24-KSC
80091-776EA 18.12 CAD
80091-776 80091-782 80091-792 80091-798 80091-804
Heavyweight Metalguard Sleeves, Wells Lamont®
Workwear
Heavyweight, cut-resistant sleeves are designed for use in applications requiring protective apparel.
  • Combination of High Performance Fiber with Stainless Steel
  • Kevlar
  • Knit Structure Allows for Air Circulation
  • Cut Resistant
  • For Use in Applications Requiring Protective Apparel
The unique knit sleeve features a patented combination of Metalguard yarn and Kevlar®.
The knit structure allows for air circulation.
Certifications: ANSI Level 4.
Caution: Caution Information: Cut-resistant sleeves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.