Identi-Plug® Plastic Foam Plugs, Jaece
Supplier: Jaece
L800-A L800-B L800-B2 L800-C L800-D L800-E L800-E2 L800-D2 L800-G
60882-145CS 251.39 CAD
60882-145 60882-167 60882-178 60882-189 60882-203 60882-205 60882-206 60882-207 89092-870
Identi-Plug® Plastic Foam Plugs, Jaece
Closures Test/Sample Tube Closures
These reusable plastic white foam plugs are for use with test tubes and flasks.
  • Provides a snug fit, yet permits free air flow
  • Proven nontoxic to algae
  • Breathable and non-absorbent
  • Autoclavable
  • Chemical resistant
  • Springs back to original shape after repeated use
These plugs can also be used as a cleaner for lab pipettes, tubes, and vials, as well as industrial precision tubing.
Ordering information: Assorted Color Variety Pack includes 150 plugs each of the following assorted colors: red, green, yellow, charcoal, blue, and white.