SAV-IT™ Closures, Porex Bio Products Group
Supplier: Thermo Fisher Scientific
A21075 A21065 A21066 A21068 A21069 A21070 A21071 A21072 A21073 A21074 A21076 A21077 A21078 A21080
28295-456CS 2324.59 CAD
28295-456 28295-458 28296-602 28296-604 28296-606 28296-608 28296-610 28296-612 28296-614 28296-616 28296-618 28296-620 28296-622 28296-624 28296-626
SAV-IT™ Closures, Porex Bio Products Group
Closures Test/Sample Tube Closures
Pliable closures for use with evacuated blood collection tubes and glass and plastic test tubes
Seals inside test tube or over tube rim. The 12mm and 13mm closures seal over the rim of either 12mm or 13mm tubes, and when reversed seal inside 10mm tubes. The 16mm closures seal over the rim of 16mm tubes, and when reversed seal inside 12mm and 13mm tubes.
They protect from contamination and prevent evaporation. Closure will not crack or pop off.
Ordering information: Supplied 1000 closures per pack, 10 packs per case.