VWR® Caps
Supplier: VWR International
60828-700 60828-702 60828-704 60828-706 60828-708 60828-720 60828-722 60828-730 60828-732 60828-738 60828-740 60828-744 60828-746 60828-748 60828-750 60828-760 60828-766 60828-772 60828-712 60828-724 60828-726 60828-728 60828-742 60828-762 60828-764 60828-768 60828-770 60828-710
60828-700PK 380.7 CAD
60828-700 60828-702 60828-704 60828-706 60828-708 60828-720 60828-722 60828-730 60828-732 60828-738 60828-740 60828-744 60828-746 60828-748 60828-750 60828-760 60828-766 60828-772 60828-712 60828-724 60828-726 60828-728 60828-742 60828-762 60828-764 60828-768 60828-770 60828-710
VWR® Caps
Closures Test/Sample Tube Closures
Safe-T-Flex caps feature a unique, flexible, over-locking design that locks securely on the outside of the tube lip and can be removed with one hand.
  • Polyethylene construction
  • Liquid-tight seal
  • Color-coded for easy specimen identification
  • Cap top can be labeled
  • Self-locking caps hold firmly in any position
A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples
Caps fit all varieties of blood collecting and disposable culture tubes. They are color-coded for easy specimen identification and to help guard againstMore Product Information