Corning® Cap, Phenolic, PTFE Liner
Supplier: Corning
9998-24 9998-13 9998-15 9998-18 9998-241
60827-225CS 109 CAD
60827-225 60827-227 60827-228 60827-229 66110-220
Corning® Cap, Phenolic, PTFE Liner
Closures Test/Sample Tube Closures
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.