DuPont™ Tyvek® 400 Sleeves
Supplier: DuPont
TY500SWH00020000 TY500SWH00020000
CA10010-136CS 410.38 CAD
CA10010-136 10010-136
DuPont™ Tyvek® 400 Sleeves
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Elastic at both ends
  • Serged seams
  • One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
These sleeves feature elastic at both biceps, measuring 8", and wrist, measuring 61/4".
Certifications: Thickness meets ASTM D1777.
Ordering information: Available standard or NAFTA sourced.