NALGENE® Screw Caps, High-Density Polyethylene, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
342151-0200 342151-0240 342151-0384
16057-316CS 81.9 CAD
16057-316 16057-318 16101-098
NALGENE® Screw Caps, High-Density Polyethylene, Sterile, Thermo Scientific
Closures Bottle Closures
For radiation-sterilization applications
Use with NALGENE® bottles. Not autoclavable.
Ordering information: Bulk packaged.
Order Now
SPECIFICATIONS
Environmentally Preferable