Nichrome Resistance Wire, Arcor Electronics
Supplier: Arcor Electronics
D14 D16 D18 D20 D22 D24 D26 D28
66258-022EA 344.2 CAD
66258-022 66258-044 66258-066 66258-088 66258-102 66258-124 66258-146 66258-168 66258-180 66258-204 66258-226
Nichrome Resistance Wire, Arcor Electronics
Wire
Used for making heating wires and inoculating loops
60% nickel, 16% chromium, 24% iron. Maximum operating temperature: 1150°C (2100°F). Bare, 1/4 lb. spool.