Sealing Disc, PTFE, Qorpak®
Supplier: Qorpak
CAP-00509 CAP-00514 CAP-00518 CAP-00522 CAP-00520 CAP-00526 CAP-00524 CAP-00528 CAP-00530 CAP-00532 CAP-00534 CAP-00537 CAP-00540 CAP-00542 CAP-00507 CAP-06829
16200-430CS 1276.62 CAD
16200-430 16200-432 16200-434 16200-436 16200-438 16200-440 16200-442 16200-444 16200-446 16200-448 16200-450 16200-452 16200-454 16200-456 16200-458 93001-504
Sealing Disc, PTFE, Qorpak®
Septa
Convert ordinary caps into non-contaminating closures at a fraction of the cost with a Polyethylene F217 Foam Disc.
  • Easy-to-clean
  • Easy to remove
  • Easy to replace
Just press the disc into your existing closure.
Order Now
SPECIFICATIONS
Environmentally Preferable