WHEATON® Aluminum Seals (Unlined), DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
224189 224176-01 224177-05 224177-06 224178-05 224193-06 W224205 W224207
10796-174CS 251.64 CAD
10796-174 10796-538 10796-540 10796-542 10796-544 10796-546 10796-628 10796-630
WHEATON® Aluminum Seals (Unlined), DWK Life Sciences
Closures Chromatography Vial Closures
Aluminum seals (unlined) are available in a wide range of styles and sizes.
  • Open top seals expose stopper for sample retrieval with a syringe
  • Tear-off seal removes completely allowing content to pour from bottle
  • Solid top seals are excellent for long-term storage of samples
  • Flip cap seals provide tamper evidence
Color selection includes natural, red, blue, and green.
Packaging: Shelf packs of 100.