KleenGuard™ A70 Chemical Spray Protection Coveralls, Kimberly-Clark Professional
Supplier: Microflex
9813 9814 9815 9816 9817 682 683 684 685 686 687
CA89030-822CS 141.59 CAD
CA89030-822 CA89030-824 CA89030-826 CA89030-828 CA89030-830 CA89030-832 CA89030-834 CA89030-836 CA89030-838 CA89030-840 CA89030-842 CA89030-844
KleenGuard™ A70 Chemical Spray Protection Coveralls, Kimberly-Clark Professional
Disposable Garments
Manufactured from NFPA 99 compliant antistatic material, coveralls feature a seamless front for superior protection, ample chest room and more body length, and a liquid-resistant, extra-long zipper
  • For use in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications
  • Features bound seams for increased protection
  • Durable, breathable, and comfortable
  • Manufactured from NFPA 99 compliant antistatic material
  • Wick-away lining provides enhanced comfort
More Product Information