White Metal Cap, Pulp/Aluminum Foil Liner, Qorpak®
Supplier: Qorpak
CAP-06652 CAP-06647 CAP-06653 CAP-06657 CAP-06648 CAP-06650 CAP-06651 CAP-06655 CAP-08684 CAP-06646 CAP-00434 CAP-00418 CAP-00439
11310-802CS 1007.97 CAD
11310-802 11310-806 11310-808 11310-810 11310-812 11310-814 11310-816 11310-818 11310-820 11311-212 11311-582 16200-400 16200-402 16200-414 11310-822 89204-922 89204-928 76535-004 76535-012 89204-923
White Metal Cap, Pulp/Aluminum Foil Liner, Qorpak®
Closures Bottle Closures
Pulp/Aluminum Foil lined caps are typically used for food applications as they will not cause odor or taste problems
They can be used for non-acid, non-alkaline products. Great for organic solvents, chrome cleaners, brake fluids and mineral oils.