White Metal Screw Caps, Plastisol Liner, Qorpak®
Supplier: Qorpak
CAP-00420 CAP-00447 CAP-00441 CAP-00442 CAP-00455 CAP-00466 CAP-00423
16151-682CS 2646.66 CAD
16151-682 16218-038 16218-118 16218-120 93001-528 16198-803 16198-825 16198-848 16198-869 16198-905 16218-041
White Metal Screw Caps, Plastisol Liner, Qorpak®
Closures Bottle Closures
White metal caps are impact-resistant and exhibit the widest range of temperature tolerances
Caps feature Plastisol liners which offer excellent resistance to mild acids. For use with glass or plastic wide mouth bottles. Not autoclavable.