VWR® Signature™ Hooded Coveralls made with DuPont™ Tyvek® IsoClean® Material
Supplier: DuPont
IC180SWHMD0025VD IC180SWHLG0025VD IC180SWH2X0025VD 89012-812 89012-814 89012-816 89012-818 89012-820
CA89012-812CS 1373.08 CAD
CA89012-812 CA89012-814 CA89012-818 89012-812 89012-814 89012-816 89012-818 89012-820
VWR® Signature™ Hooded Coveralls made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Coveralls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms.
  • Protective, durable, and comfortable
  • Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Chemically and biologically inert, low linting, and static free until laundered
  • Serged seams, front zipper closure, elastic wrists, and attached hood
  • Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles
Antistatic coveralls are low-linting and chemically and biologically inert.
All coveralls feature serged seams, front zipper closure, elastic wrists, and attached hood. Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles.