Nalgene® Barbed Bulkhead Fittings, Thermo Scientific
Supplier: Thermo Fisher Scientific
6149-0001 6149-0002
16225-232CS 611.82 CAD
16225-232 16331-102
Nalgene® Barbed Bulkhead Fittings, Thermo Scientific
Fittings and Connectors
All-plastic autoclavable fitting permits retrofitting of most NALGENE® and other manufacturers’ closures for liquid transfer when venting is not required
Unique fitting with two barbed ends is useful wherever tubing must be attached on both ends. Gasket and nut assemblies provide leakproof service. Can be sterilized by autoclave, gas or chemical methods. For vacuum applications, use with NALGENE® heavy-duty 4L bottle, 16331-100, with an 83mm cap. Use with 53mm cap for liquid transfer on 1L and 2L HDPE bottles or 8L bottles.
Ordering information: Complete with instructions and template for installation.