NALGENE® High-Density Polyethylene Screw Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
712151-0053 712151-0200 712151-0380 712151-0083
16219-108PK 261.57 CAD
16219-108 16219-112 16219-116 16225-162
NALGENE® High-Density Polyethylene Screw Closures, Thermo Scientific
Closures Bottle Closures
For radiation-sterilization applications
Use with NALGENE® bottles. Not autoclavable.