DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Bound Seams
Supplier: DuPont
Tychem®
QC125BYL6X001200 QC125BYL5X001200 QC125BYL3X00 QC125BYLMD00 QC125BYL3X00 QC125BYL2X00 QC125BYLXL00 QC125BYL4X00 QC125BYLXL00 QC125BYLMD00 QC125BYL4X00 QC125BYL2X00 QC125BYLLG00 QC125BYLLG00
10016-396CS 156.84 CAD
13452-638 13452-636 13452-646 13452-644 13452-642 13452-640 10016-396 10016-398 CA13452-640 CA13452-642 CA13452-644 CA13452-646 CA13452-636 CA13452-638
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Laydown collar
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewn seams are covered with garment fabric to reinforce seam and reduce potential for particle penetration. Coveralls feature a laydown collar, elastic wrists, and elastic ankles. Storm flap covers zipper, which can be sealed by the wearer with an adhesive strip to prevent intrusion at zipper.
Certifications: Thickness meets ASTM D1777.