NALGENE® Polypropylene Screw Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
712150-0240 712150-0530 712150-0430 712150-0630 712150-1000 712174-0530 712171-0530 712150-0280
16219-030PK 43.88 CAD
16219-030 16219-080 16219-102 16219-104 16219-106 16219-150 16219-301 16219-041
NALGENE® Polypropylene Screw Closures, Thermo Scientific
Closures Bottle Closures
Linerless closures are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable. Available in natural and amber.