PMC Series Plastic Couplings, Colder Products Company
Supplier: Colder Products
PMC1604 PMC4204 PMCD4204 PMC2202
16001-808PK 328.1 CAD
16001-808 16001-816 16001-822 16001-828 16001-832 16001-836 16001-842 16001-860 16001-872 16001-876 16001-880 16001-882 16001-886 16001-888 16001-892 16001-898 16001-904 16001-912 16001-918 16001-926 16001-932 16001-940 16001-944 16001-954 16001-972 16001-976 16001-980 16001-988 16001-854 16001-960
PMC Series Plastic Couplings, Colder Products Company
Fittings and Connectors
These quick disconnect couplings cover a wide range of general purpose applications from chemical analyzers to medical devices
They feature integral terminations, which results in fewer leak points, shorter assemblies, and faster installations. An audible click assures the user of a secure connection. Couplings include a thumb latch for one-hand connection and disconnection without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues. Components and valves are constructed of acetal thermoplastic, with stainless steel latches, springs, and pins. O-rings are made of Buna-N. Couplings are produced in a natural white tint.
Ordering information: Polypropylene couplings for greater chemical resistance are also available; contact your Avantor sales representative.