PLC Series Plastic Couplings, Colder Products Company
Supplier: Colder Products
PLC12004 PLC13004 PLC13006 PLC40004 PLCD40004 PLCD42006
16001-642PK 256.02 CAD
16001-642 16001-644 16001-648 16001-650 16001-654 16001-660 16001-664 16001-668 16001-672 16001-674 16001-678 16001-684 16001-696 16001-700 16001-706 16001-708 16001-712 16001-714 16001-718 16001-724 16001-726 16001-730 16001-740 16001-746 16001-750 16001-756 16001-760 16001-762 16001-768 16001-774 16001-786 16001-790 16001-796 16001-806
PLC Series Plastic Couplings, Colder Products Company
Fittings and Connectors
These quick disconnect couplings are injection molded from acetal thermoplastic, making them highly durable and resistant to most chemical solutions
Both the external and valve springs are constructed of stainless steel. Couplings feature integral terminations which results in fewer leak points, shorter assemblies, and faster installations. An audible click assures the user of a secure and reliable connection. Couplings have a thumb latch for rapid disconnect and reconnect with one hand, without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues.
Ordering information: Polypropylene couplings for greater chemical resistance are available; contact your Avantor sales representative.