DuPont™ Tychem® 4000 Coveralls with Standard Hood, Bound Seams
Supplier: DuPont
SL127BWHMD001200 SL127BWHLG001200 SL127BWHXL001200 SL127BWH2X001200 SL127BWH3X001200 SL127BWH4X001200 SL127BWH5X001200 SL127BWH6X001200 SL127BWH7X001200
CA10010-168CS 2018.67 CAD
CA10010-168 CA10010-170 CA10010-172 CA10010-174 CA10010-176 CA10010-178 10016-490 10016-492 10016-494
DuPont™ Tychem® 4000 Coveralls with Standard Hood, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Tightly sewnMore Product Information