Synthware Anti-Splash Adapters with Fritted Disc
Supplier: Kemtech America
A232414 A232419 A232424 A231250 A234250
60000-432EA 269.31 CAD
60000-432 60000-434 60000-436 60000-446 60000-450
Synthware Anti-Splash Adapters with Fritted Disc
Adapters Adapters, Conical Ground Joints
Anti-splash adapters prevent possible contamination from evaporator.
  • Extra coarse fritted disc
  • Top [ST] joint
  • Bottom [ST] joint
Adapters feature an extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from carrying over.