Synthware Anti-Climb Adapters
Supplier: Kemtech America
A272414 A272419 A274100 A272250 A274250 A279450
60000-378EA 236.68 CAD
60000-378 60000-380 60000-382 60000-390 60000-394 60000-400
Synthware Anti-Climb Adapters
Adapters Adapters, Conical Ground Joints
The trap of the anti-climb adapter provides expanded volume for reactions that have a tendency to foam excessively.
  • Sealed inner vapor tube
  • Top [ST] joint
  • Bottom [ST] joint
Adapter features an inner vapor tube that is sealed to the top of the trap to prevent entry into the condenser.