Bushing Adapters, Ace Glass Incorporated
Supplier: Ace Glass
5021-05 5021-07 5021-09 5021-12 5021-18 5021-20 5021-24 5021-26 5021-28 5021-30 5021-35 5021-36 5021-39 5021-94 9061-05 9061-10 9061-16 5021-14 5021-15 5021-22
89051-044EA 123.44 CAD
89051-044 89051-046 89051-048 89051-050 89051-052 89051-054 89051-056 89051-058 89051-060 89051-062 89051-064 89051-066 89051-068 89051-070 89064-812 89064-814 89064-816 89082-608 89082-610 89082-612
Bushing Adapters, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These bushing-type reducing adapters are manufactured from borosilicate glass.
  • Larger bottom outside [ST] joint
  • Smaller top inside [ST] joint
Adapters feature a larger bottom outside standard taper joint, and a smaller top inside standard taper joint.