Polypropylene Coveralls, International ENVIROGUARD™
Supplier: International Enviroguard
2012-L 2012-XL 2012-4XL N2012-L N2012-XL N2012-3XL
83007-016CS 122.88 CAD
83007-016 83007-018 83007-020 83007-022 83007-024 83007-026 83007-028 83007-032
Polypropylene Coveralls, International ENVIROGUARD™
Disposable Garments
Disposable coveralls protect against dirt, grime, and certain dry particulates in nonhazardous environments
They are ideal for use in industrial facilities, pharmaceutical applications, food processing, and certain environmental cleanup sites. Coveralls have elastic wrists and open ankles.