Nalgene® Serum Vial Closures, HDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
342158-0021 342158-0023 342158-0026 342158-0024 342158-0025 312158-0021
16060-100CS 1184.37 CAD
16060-100 73520-090 73520-092 73520-094 73520-096 16059-992
Nalgene® Serum Vial Closures, HDPE, Thermo Scientific
Closures Chromatography Vial Closures
These 20mm, high-density polyethylene screw cap closures are available lined or unlined and sterile or nonsterile.
Ordering information: All closures are bulk packed. Sterile closures are bulk packed in a case with a three-ply liner. Unlined closures require a stopper (not included).