DuPont™ Tyvek® IsoClean® Coveralls with Standard Collar
Supplier: DuPont
IC181SWHSM002500 IC181SWHXL002500 IC181SWHSM00250C IC181SWH4X00250C IC181SWH5X002500 IC181SWH6X002500 IC181SWH7X002500
CA80076-744CS 2115.54 CAD
CA80076-744 CA80076-746 CA80076-750 CA80076-786 CA80076-948 CA80077-288 CA80076-748 CA80076-752 10016-064 10016-066 10016-068 10016-070
DuPont™ Tyvek® IsoClean® Coveralls with Standard Collar
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Standard laydown collar
  • Set sleeve design
  • Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Collar and closure allow for easy donning andMore Product Information